Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-23
Entrez Gene ID
185369
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_F36G9.6
Chromosome Location
Chr.V: 15970376 - 15972584 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074603.3 NP_507004.2 4 - 5 802 97

View Full Product Details