Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-44
Entrez Gene ID
188375
Gene Name
7TM_GPCR_Srx domain-containing protein
Gene Aliases
CELE_T10C6.4
Chromosome Location
Chr.V: 16020342 - 16022652 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074622.4 NP_507023.3 6 - 7 753 99

View Full Product Details