Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srxa-16
Entrez Gene ID
185755
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F44G3.5
Chromosome Location
Chr.V: 16112992 - 16114337 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074655.2 NP_507056.2 3 - 4 498 154

View Full Product Details