Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpl-1
Entrez Gene ID
180111
Gene Name
protein-coding
Gene Aliases
CELE_T03E6.7
Chromosome Location
Chr.V: 16584168 - 16595348 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269789.3 NP_001256718.1 3 - 4 812 71
NM_001269790.3 NP_001256719.1 1 - 2 392 71

View Full Product Details