Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-43
Entrez Gene ID
184479
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F14F8.5
Chromosome Location
Chr.V: 16686025 - 16687083 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074824.2 NP_507225.2 1 - 2 75 166

View Full Product Details