Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
phy-3
Entrez Gene ID
188624
Gene Name
Fe2OG dioxygenase domain-containing protein
Gene Aliases
CELE_T20B3.7
Chromosome Location
Chr.V: 16833165 - 16839141 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074850.3 NP_507251.2 4 - 5 732 90

View Full Product Details