Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-95
Entrez Gene ID
190855
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_Y102A5C.33
Chromosome Location
Chr.V: 16992034 - 16993910 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074898.1 NP_507299.1 4 - 5 734 138

View Full Product Details