Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nhr-65
Entrez Gene ID
180230
Gene Name
protein-coding
Gene Aliases
CELE_Y17D7A.3
Chromosome Location
Chr.V: 18839304 - 18845447 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029048.6 NP_001024219.1 1 - 2 125 111
NM_001029049.6 NP_001024220.1 1 - 2 131 111

View Full Product Details