Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-79
Entrez Gene ID
180265
Gene Name
protein-coding
Gene Aliases
CELE_T26H2.9
Chromosome Location
Chr.V: 19207676 - 19217180 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373193.1 NP_001360177.1 6 - 7 697 76
NM_075333.8 NP_507734.4 10 - 11 1491 76

View Full Product Details