Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ttr-13
- Entrez Gene ID
-
189897
- Gene Name
- TransThyretin-Related family domain
- Gene Aliases
-
CELE_Y44A6B.4
- Chromosome Location
-
Chr.V: 20635901 - 20636955 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075585.3 |
NP_507986.1 |
3 - 4 |
416 |
104 |
- Gene Symbol
- Y44A6B.3
- Entrez Gene ID
-
189896
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_Y44A6B.3
- Chromosome Location
-
Chr.V: 20635891 - 20638825 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_075586.3 |
NP_507987.2 |
5 - 6 |
1623 |
104 |
View Full Product Details