Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-35
Entrez Gene ID
186180
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F53F8.2
Chromosome Location
Chr.V: 20681420 - 20683014 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075593.2 NP_507994.2 4 - 5 439 126

View Full Product Details