Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arr-1
Entrez Gene ID
180446
Gene Name
Beta-arrestin arr-1
Gene Aliases
CELE_F53H8.2
Chromosome Location
Chr.X: 950773 - 954233 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075782.7 NP_508183.1 2 - 3 157 67

View Full Product Details