Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snx-1
Entrez Gene ID
180468
Gene Name
PX domain-containing protein
Gene Aliases
CELE_C05D9.1
Chromosome Location
Chr.X: 1128984 - 1133431 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075815.7 NP_508216.1 2 - 3 374 84

View Full Product Details