Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hum-6
Entrez Gene ID
180539
Gene Name
Unconventional myosin heavy chain 6
Gene Aliases
CELE_T10H10.1
Chromosome Location
Chr.X: 2303078 - 2314914 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076019.5 NP_508420.1 3 - 4 319 157

View Full Product Details