Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-16
Entrez Gene ID
180594
Gene Name
TWiK family of potassium channels
Gene Aliases
CELE_F52E4.4
Chromosome Location
Chr.X: 3081125 - 3085401 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076125.4 NP_508526.2 4 - 5 373 150
GenBank mRNA AF083645.1 - 4 - 5 365 150

View Full Product Details