Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cht-1
Entrez Gene ID
180628
Gene Name
Probable endochitinase
Gene Aliases
CELE_C04F6.3
Chromosome Location
Chr.X: 3397613 - 3400017 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076187.5 NP_508588.1 1 - 2 87 58

View Full Product Details