Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npr-1
Entrez Gene ID
180752
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C39E6.6
Chromosome Location
Chr.X: 4767152 - 4769985 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076415.6 NP_508816.1 7 - 8 1107 94

View Full Product Details