Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-48
Entrez Gene ID
188031
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_T03G11.2
Chromosome Location
Chr.X: 5180174 - 5181495 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076480.2 NP_508881.2 1 - 2 231 91

View Full Product Details