Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lite-1
Entrez Gene ID
180894
Gene Name
High-energy light unresponsive protein 1
Gene Aliases
CELE_C14F11.3
Chromosome Location
Chr.X: 6226126 - 6230769 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076642.7 NP_509043.3 5 - 6 634 131

View Full Product Details