Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
atg-2
Entrez Gene ID
180949
Gene Name
Autophagy-related protein 2
Gene Aliases
CELE_M03A8.2
Chromosome Location
Chr.X: 6806729 - 6817266 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076744.9 NP_509145.3 6 - 7 1290 144

View Full Product Details