Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pes-23
Entrez Gene ID
180963
Gene Name
MFS domain-containing protein
Gene Aliases
CELE_F14B8.3
Chromosome Location
Chr.X: 6901378 - 6903986 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076766.9 NP_509167.2 1 - 2 224 90

View Full Product Details