Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
otpl-3
Entrez Gene ID
259710
Gene Name
OToPetrin-Like
Gene Aliases
CELE_F14B8.7
Chromosome Location
Chr.X: 6921646 - 6924764 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076769.4 NP_509170.1 10 - 11 1492 75

View Full Product Details