Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
alh-10
Entrez Gene ID
180979
Gene Name
Aldedh domain-containing protein
Gene Aliases
CELE_C54D1.4
Chromosome Location
Chr.X: 7140504 - 7143641 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076802.5 NP_509203.1 4 - 5 950 112

View Full Product Details