Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lam-2
Entrez Gene ID
180980
Gene Name
Laminin-like protein lam-2
Gene Aliases
CELE_C54D1.5
Chromosome Location
Chr.X: 7143846 - 7151239 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076803.7 NP_509204.3 1 - 2 347 86

View Full Product Details