Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hlh-8
Entrez Gene ID
181069
Gene Name
Twist-related protein
Gene Aliases
CELE_C02B8.4
Chromosome Location
Chr.X: 8116457 - 8120340 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076966.3 NP_509367.1 4 - 5 463 66
GenBank mRNA AF037063.1 - 3 - 4 368 66

View Full Product Details