Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbc-18
Entrez Gene ID
181091
Gene Name
TBC (Tre-2/Bub2/Cdc16) domain family
Gene Aliases
CELE_F41D9.1
Chromosome Location
Chr.X: 8378557 - 8382711 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077020.7 NP_509421.2 2 - 3 156 81

View Full Product Details