Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ced-8
Entrez Gene ID
184190
Gene Name
Cell death abnormality protein 8
Gene Aliases
CELE_F08F1.5
Chromosome Location
Chr.X: 8414006 - 8416919 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077026.10 NP_509427.2 2 - 3 333 103

View Full Product Details