Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dylt-2
Entrez Gene ID
183892
Gene Name
DYnein Light chain (Tctex type)
Gene Aliases
CELE_D1009.5
Chromosome Location
Chr.X: 8939220 - 8940159 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077110.3 NP_509511.1 1 - 2 73 80

View Full Product Details