The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001375094.1 | NP_001362077.1 | 1 - 2 | 88 | 124 |
NM_077145.7 | NP_509546.4 | 1 - 2 | 88 | 124 |