Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
flp-20
Entrez Gene ID
181165
Gene Name
FMRF-Like Peptide
Gene Aliases
CELE_E01H11.3
Chromosome Location
Chr.X: 9349276 - 9349894 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077173.6 NP_509574.2 1 - 2 136 74

View Full Product Details