Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-41
Entrez Gene ID
183331
Gene Name
protein-coding
Gene Aliases
CELE_C39B10.2
Chromosome Location
Chr.X: 10454260 - 10460506 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129644.2 NP_001123116.1 5 - 6 515 82
NM_001373473.2 NP_001360683.1 5 - 6 515 82
NM_001373474.1 NP_001359532.1 5 - 6 515 82
NM_001373475.2 NP_001360682.1 5 - 6 516 82
NM_001373476.2 NP_001360686.1 5 - 6 515 82
NM_001373477.1 NP_001360687.1 5 - 6 515 82
NM_001373478.3 NP_001360684.1 6 - 6 558 82
NM_001373479.2 NP_001360685.1 6 - 6 563 82
NM_001373480.3 NP_001360688.1 6 - 6 564 82
NM_001373481.2 NP_001360689.1 6 - 6 554 82

View Full Product Details