Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
elt-2
Entrez Gene ID
181250
Gene Name
Transcription factor elt-2
Gene Aliases
CELE_C33D3.1
Chromosome Location
Chr.X: 10481240 - 10483446 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077354.5 NP_509755.2 3 - 4 370 120

View Full Product Details