Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
git-1
Entrez Gene ID
181253
Gene Name
GIT1 (mammalian G protein-coupled receptor kinase InTeractor 1) homolog
Gene Aliases
CELE_F14F3.2
Chromosome Location
Chr.X: 10524898 - 10528469 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077360.8 NP_509761.2 8 - 9 1156 97

View Full Product Details