Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
syd-2
Entrez Gene ID
181255
Gene Name
Liprin-alpha
Gene Aliases
CELE_F59F5.6
Chromosome Location
Chr.X: 10549100 - 10555241 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077367.9 NP_509768.1 5 - 6 756 78
GenBank mRNA AF170122.1 - 4 - 5 683 78

View Full Product Details