Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acbp-3
Entrez Gene ID
181281
Gene Name
Acyl-CoA-binding protein homolog 3
Gene Aliases
CELE_F47B10.7
Chromosome Location
Chr.X: 10903278 - 10905107 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077421.6 NP_509822.2 2 - 3 189 69

View Full Product Details