Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
alr-1
Entrez Gene ID
181302
Gene Name
Homeobox domain-containing protein
Gene Aliases
CELE_R08B4.2
Chromosome Location
Chr.X: 11121871 - 11125620 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077459.6 NP_509860.1 3 - 4 502 62

View Full Product Details