Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slo-2
Entrez Gene ID
181332
Gene Name
protein-coding
Gene Aliases
CELE_F08B12.3
Chromosome Location
Chr.X: 11394605 - 11405931 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029356.4 NP_001024527.1 17 - 18 3428 152
NM_001136452.4 NP_001129924.1 17 - 18 3112 152
GenBank mRNA AF173828.1 - 17 - 18 3320 152

View Full Product Details