Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srd-50
Entrez Gene ID
184507
Gene Name
Serpentine receptor class delta-50
Gene Aliases
CELE_F15A2.4
Chromosome Location
Chr.X: 13478621 - 13481804 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322645.3 NP_001309446.1 2 - 3 439 113

View Full Product Details