Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ztf-14
Entrez Gene ID
187492
Gene Name
protein-coding
Gene Aliases
CELE_M163.2
Chromosome Location
Chr.X: 14496085 - 14499436 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373576.2 NP_001360586.1 2 - 3 452 90
NM_001373577.2 NP_001360587.1 2 - 2 417 90

View Full Product Details