Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lea-1
Entrez Gene ID
3564838
Gene Name
protein-coding
Gene Aliases
CELE_K08H10.1
Chromosome Location
Chr.V: 10011923 - 10028168 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269231.3 NP_001256160.1 11 - 12 4084 87
NM_001269232.3 NP_001256161.1 9 - 10 3535 87
NM_001269234.3 NP_001256163.1 10 - 11 4036 87
NM_001269235.3 NP_001256164.1 8 - 9 3487 87
NM_001269240.3 NP_001256169.1 9 - 10 3820 87
NM_001269241.4 NP_001256170.1 7 - 8 3271 87
NM_001269244.3 NP_001256173.1 4 - 5 1561 87
NM_001269245.3 NP_001256174.1 3 - 4 1384 87
NM_001373085.2 NP_001360513.1 10 - 10 3907 87
NM_001373086.2 NP_001360514.1 10 - 10 3712 87
NM_001373087.2 NP_001360515.1 9 - 9 3859 87
NM_001373088.2 NP_001360516.1 9 - 10 3664 87
NM_001373089.2 NP_001360517.1 8 - 8 3643 87
NM_001373090.2 NP_001360518.1 8 - 8 3448 87

View Full Product Details