Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-21
Entrez Gene ID
183488
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_C45H4.7
Chromosome Location
Chr.V: 2155532 - 2156635 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071183.2 NP_503584.2 3 - 4 514 116

View Full Product Details