Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gsp-2
Entrez Gene ID
3564807
Gene Name
Serine/threonine-protein phosphatase PP1-beta
Gene Aliases
CELE_F56C9.1
Chromosome Location
Chr.III: 7336587 - 7338769 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027445.6 NP_001022616.1 6 - 7 917 82

View Full Product Details