Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB33A
Entrez Gene ID
612988
Gene Name
RAB33A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.X: 101311752 - 101322825 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022416056.1 XP_022271764.1 2 - 3 525 69

View Full Product Details