Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL21
Entrez Gene ID
442935
Gene Name
interleukin 21
Gene Aliases
IL-21
Chromosome Location
Chr.19: 17641833 - 17651275 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003347.1 NP_001003347.1 - 252 100
GenBank mRNA AB127975.1 - - 252 100

View Full Product Details