Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CES5A
Entrez Gene ID
445456
Gene Name
carboxylesterase 5A
Gene Aliases
CES7
Chromosome Location
Chr.2: 60235986 - 60265584 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003969.1 NP_001003969.1 2 - 3 282 56
XM_022405449.1 XP_022261157.1 2 - 3 476 56
GenBank mRNA AB186392.1 - 2 - 3 282 56

View Full Product Details