Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MS4A1
Entrez Gene ID
485430
Gene Name
membrane spanning 4-domains A1
Gene Aliases
CD20
Chromosome Location
Chr.21: 50722142 - 50734934 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048028.1 NP_001041493.1 2 - 3 206 75
XM_005633300.3 XP_005633357.1 2 - 3 332 75
XM_022407438.1 XP_022263146.1 2 - 3 251 75
GenBank mRNA AB210085.1 - 2 - 3 206 75

View Full Product Details