Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2E1
Entrez Gene ID
415128
Gene Name
cytochrome P450 family 2 subfamily E member 1
Gene Aliases
-
Chromosome Location
Chr.28: 41079164 - 41089188 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003339.1 NP_001003339.1 7 - 8 1159 66
GenBank mRNA AF029978.1 - 7 - 8 1157 66
AF029979.1 - 7 - 8 1159 66

View Full Product Details