Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFF1
Entrez Gene ID
403491
Gene Name
trefoil factor 1
Gene Aliases
-
Chromosome Location
Chr.31: 36681478 - 36684571 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002992.1 NP_001002992.1 1 - 2 104 116
GenBank mRNA AY258287.1 - 1 - 2 104 116
EF591857.1 - 1 - 2 98 116
EF591858.1 - 1 - 2 98 116
EF591859.1 - 1 - 2 99 116

View Full Product Details