Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMN
Entrez Gene ID
403896
Gene Name
survival motor neuron
Gene Aliases
-
Chromosome Location
Chr.2: 54596000 - 54636837 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003226.2 NP_001003226.1 8 - 9 873 82
XM_022405358.1 XP_022261066.1 8 - 9 918 82
GenBank mRNA AY289762.1 - 8 - 9 859 82
U50746.1 - 8 - 9 874 82

View Full Product Details