Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESRRA
Entrez Gene ID
403169
Gene Name
estrogen related receptor alpha
Gene Aliases
ESTRRA
Chromosome Location
Chr.18: 52755547 - 52762938 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002936.1 NP_001002936.1 5 - 6 1002 81
XM_005631023.2 XP_005631080.1 6 - 7 1158 81
XM_022405056.1 XP_022260764.1 6 - 7 1406 81
GenBank mRNA AY533243.1 - 5 - 6 1002 81

View Full Product Details